Doxycycline hyclate, 98%
Doxycycline hyclate, 98%
Doxycycline hyclate, 98%
Doxycycline hyclate, 98%
Thermo Scientific Chemicals
Doxycycline hyclate, 98%
Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number FSA446061000
Price (MYR)
1,997.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,997.00
EA
Add to cart
Specifications
Chemical Name or MaterialDoxycycline hyclate
CAS24390-14-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
View more
General description
  • Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
  • Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
  • Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
  • Inhibits matrix metalloproteinases in many microorganisms
Applications
  • Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
  • Intended for laboratory use only
RUO – Research Use Only