Tetramethylsilane, ACS reagent, NMR grade
Tetramethylsilane, ACS reagent, NMR grade
Tetramethylsilane, ACS reagent, NMR grade
Tetramethylsilane, ACS reagent, NMR grade
Thermo Scientific Chemicals
Tetramethylsilane, ACS reagent, NMR grade
CAS: 75-76-3 | C4H12Si | 88.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA445901000
Price (MYR)
614.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
614.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylsilane
CAS75-76-3
Health Hazard 1GHS Signal Word: Danger
Melting Point-99°C