Paraffin, liquid, technical
Paraffin, liquid, technical
Paraffin, liquid, technical
Paraffin, liquid, technical
Thermo Scientific Chemicals
Paraffin, liquid, technical
CAS: 8012-95-1 | MFCD00131611
Have Questions?
Catalog NumberQuantityPackaging
FSA44418025025 LPlastic Can
Catalog number FSA444180250
Price (MYR)
2,126.00
EA
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
2,126.00
EA
Add to cart
Specifications
Chemical Name or MaterialParaffin, liquid
CAS8012-95-1
Health Hazard 1GHS Signal Word: Warning
Melting Point-24.0°C
Density0.8800g/mL