2-Amino-6-bromobenzothiazole, 95%
2-Amino-6-bromobenzothiazole, 95%
2-Amino-6-bromobenzothiazole, 95%
2-Amino-6-bromobenzothiazole, 95%
Thermo Scientific Chemicals
2-Amino-6-bromobenzothiazole, 95%
CAS: 15864-32-1 | C7H5BrN2S | 229.10 g/mol
Have Questions?
Catalog NumberQuantity
FSA4422900505 g
Catalog number FSA442290050
Price (MYR)
889.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
889.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-6-bromobenzothiazole
CAS15864-32-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0