3-Fluoro-4-pyridinecarboxaldehyde, 97%
3-Fluoro-4-pyridinecarboxaldehyde, 97%
3-Fluoro-4-pyridinecarboxaldehyde, 97%
3-Fluoro-4-pyridinecarboxaldehyde, 97%
Thermo Scientific Chemicals
3-Fluoro-4-pyridinecarboxaldehyde, 97%
CAS: 40273-47-0 | C6H4FNO | 125.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA442170010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Fluoro-4-pyridinecarboxaldehyde
CAS40273-47-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash