Check Order Status
2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, Thermo Scientific™
Element Search
Structure Search
2-Fluoro-6-(trifluoromethyl)benzyl bromide, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Catalog number
FSA442160010
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
2-Fluoro-6-(trifluoromethyl)benzyl bromide
CAS
239087-08-2
Health Hazard 1
GHS Signal Word:
Danger
Melting Point
42°C to 44°C
Flash Point
107°C