2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA441911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Phenylethylamine hydrochloride
CAS156-28-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0