Boron nitride, 98%
Boron nitride, 98%
Boron nitride, 98%
Thermo Scientific Chemicals
Boron nitride, 98%
CAS: 10043-11-5 | BN | 24.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA441800500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron nitride
Melting Point2700.0°C to 3000.0°C
CAS10043-11-5
Quantity50 g
EINECS Number233-136-6