Strontium nitrate, 98%, extra pure
Strontium nitrate, 98%, extra pure
Strontium nitrate, 98%, extra pure
Strontium nitrate, 98%, extra pure
Thermo Scientific Chemicals
Strontium nitrate, 98%, extra pure
CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA438520010
Price (MYR)
729.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
729.00
EA
Add to cart
Specifications
Chemical Name or MaterialStrontium nitrate
Melting Point570.0°C
Name NoteExtra Pure
CAS10042-76-9