Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)
CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA437200050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen hexachloroplatinate hydrate
Melting Point60.0°C
CAS26023-84-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
View more
For ultra trace analysis
RUO – Research Use Only