Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA436961000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point210.0°C
Chemical Name or MaterialAmmonium nitrate
Melting Point169.0°C
CAS6484-52-2
ColorWhite
View more
For ultra trace analysis
RUO – Research Use Only