Molecular sieves 4A, Cylinders, 3.2mm diameter
Molecular sieves 4A, Cylinders, 3.2mm diameter
Molecular sieves 4A, Cylinders, 3.2mm diameter
Molecular sieves 4A, Cylinders, 3.2mm diameter
Thermo Scientific Chemicals
Molecular sieves 4A, Cylinders, 3.2mm diameter
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number FSA436730010
Price (MYR)
625.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
625.00
EA
Add to cart
Specifications
Bulk Density650 to 750g/L
Chemical Name or MaterialMolecular sieves 4A
ColorBeige
PackagingGlass Bottle
Residual Water< 2% (900°C, 2 hrs.)
View more
Drying agent
RUO – Research Use Only