β-Estradiol, 98%
β-Estradiol, 98%
β-Estradiol, 98%
Thermo Scientific Chemicals
β-Estradiol, 98%
β-Estradiol, 98%, CAS # 50-28-2, is an estrogen steroid sex hormone that regulates several biological functions including gene transcription and protein synthesis.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA436320250
Price (MYR)
3,065.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,065.00
EA
Add to cart
Specifications
Chemical Name or Materialβ-Estradiol
CAS50-28-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
General Description
• β-Estradiol is a naturally occurring endogenous hormone in females
• It is an important steroid sex hormone involved in the regulation of several biological functions including gene transcription and protein synthesis
• This hormone is produced by the ovaries and can bind to and activate specific nuclear receptors
Applications
• β-Estradiol acts as an estrogenic hormone receptor agonist and activator of PI 3-kinase
• It shows inhibitory activity against aldehyde oxidase
• β-Estradiol presents mild anabolic and metabolic activities and increases blood coagulability in vivo experiments
• It plays a role in immune and inflammatory processes
• β-Estradiol triggers an increase in hepatic synthesis of some proteins, including sex hormone-binding globulin (SHBG), and thyroid-binding globulin (TBG), and suppresses the formation of follicle-stimulating hormone (FSH)
• This compound has been used for the in vitro maturation of bovine cumulus-oocyte complexes (COCs)
RUO – Research Use Only
General References:
  1. Beker, A.R.; Colenbrander, B.; Bevers, M.M. Effect of 17beta-estradiol on the in vitro maturation of bovine oocytes. Theriogenology. 2002, 58, (9), 1663-1673.
  2. Maksura, H.; Akon, N.; Islam, M.N.; Akter, I.; Modak, A.K.; Khatun, A.; Alam, M.H.; Hashem, M.A.; Amin, M.R.; Moniruzzaman, M. Effects of estradiol on in vitro maturation of buffalo and goat oocytes. Reprod Med Biol. 2020 Sep 25;20(1):62-70.
  3. Liu, X.; Liu, Y.; Cheng, M.; Zhang, X.; Xiao, H. A metabolomics study of the inhibitory effect of 17-beta-estradiol on osteoclast proliferation and differentiation. Mol. Biosyst. 2015, 11 (2), 635-646.
  4. Pooley, A. E.; Luong, M.; Hussain, A.; Nathan, B. P. Neurite outgrowth promoting effect of 17-beta estradiol is mediated through estrogen receptor alpha in an olfactory epithelium culture. Brain Res. 2015, 1624, 19-27.