Kaempferol, 97%
Kaempferol, 97%
Kaempferol, 97%
Thermo Scientific Chemicals
Kaempferol, 97%
CAS: 520-18-3 | C15H10O6 | 286.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA434731000100 mgGlass bottle
Catalog number FSA434731000
Price (MYR)
2,845.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,845.00
EA
Add to cart
Specifications
Chemical Name or MaterialKaempferol
CAS520-18-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.