Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Thermo Scientific Chemicals
Poly(ethylene glycol), average M.W. 4000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA434630010
Price (MYR)
483.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
483.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene glycol)
ColorWhite
CAS25322-68-3
Physical FormFlakes
Melting Point54.0°C to 58.0°C