n-Butyl acetate, ACS reagent
n-Butyl acetate, ACS reagent
n-Butyl acetate, ACS reagent
n-Butyl acetate, ACS reagent
Thermo Scientific Chemicals
n-Butyl acetate, ACS reagent
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA434621000
Price (MYR)
252.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
252.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Butyl acetate
CAS123-86-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.