1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
1-Octanol, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
1-Octanol, 99%, anhydrous, AcroSeal™
CAS: 111-87-5 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA434581000
Price (MYR)
438.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
438.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octanol
CAS111-87-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0