Tris(hydroxymethyl)aminomethane acetate salt, 99%
Tris(hydroxymethyl)aminomethane acetate salt, 99%
Tris(hydroxymethyl)aminomethane acetate salt, 99%
Tris(hydroxymethyl)aminomethane acetate salt, 99%
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane acetate salt, 99%
CAS: 6850-28-8 | C4H11NO3·C2H4O2 | 181.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA434500250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTris(hydroxymethyl)aminomethane acetate salt
CAS6850-28-8
Absorbance0.5 max. (c=50, water) at 290nm
ColorWhite
EINECS Number229-939-6
View more
Biological buffer
RUO – Research Use Only