Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
CAS: 429-41-4 | C16H36FN | 261.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number FSA433518000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS429-41-4, 109-99-9, 7732-18-5
Specifications
Chemical Name or MaterialTetrabutylammonium Fluoride
CAS429-41-4
ColorBrown to Green
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
May cause drowsiness or dizzines