Dichlorodimethylsilane, 99+%, AcroSeal™
Dichlorodimethylsilane, 99+%, AcroSeal™
Dichlorodimethylsilane, 99+%, AcroSeal™
Dichlorodimethylsilane, 99+%, AcroSeal™
Thermo Scientific Chemicals
Dichlorodimethylsilane, 99+%, AcroSeal™
CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA430888000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDichlorodimethylsilane
CAS75-78-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p