Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Dimethyl sulfate, 99+%, AcroSeal™
Thermo Scientific Chemicals
Dimethyl sulfate, 99+%, AcroSeal™
CAS: 77-78-1 | C2H6O4S | 126.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4308300101 LAcroSeal™ Glass Bottle
Catalog number FSA430830010
Price (MYR)
514.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
514.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl sulfate
CAS77-78-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory i
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
View more
Alkylation reagent
RUO – Research Use Only