Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
Thermo Scientific Chemicals
Hydrogen bromide, pure, 33 wt.% sol. in glacial acetic acid, AcroSeal™
CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA430698000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen bromide
CAS10035-10-6
ColorYellow
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.