Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
CAS: 122-51-0 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA429271000
Price (MYR)
456.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
456.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriethyl orthoformate
CAS122-51-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.