Titanium(III) chloride, 20%w/v solution in 2N Hydrochloric acid,AcroSeal™
CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FSA429151000
100 mL
AcroSeal™ Glass Bottle
Catalog number FSA429151000
Price (MYR)
649.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
649.00
EA
Add to cart
Chemical Identifiers
CAS7705-07-9, 7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialTitanium(III) chloride
CAS7705-07-9
ColorPurple
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage. May be corrosive to metals.