Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Thermo Scientific Chemicals
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA428958000
Price (MYR)
2,772.00
EA
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,772.00
EA
Add to cart
Chemical Identifiers
CAS2052-49-5, 67-56-1
Specifications
Boiling Point65.0°C
Chemical Name or MaterialTetrabutylammonium Hydroxide
Melting Point-98.0°C
CAS2052-49-5
ColorColorless to Yellow