Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
Iodine monobromide, 1M solution in methylene chloride, AcroSeal™
CAS: 7789-33-5 | BrI | 206.81 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA428701000100 mLAcroSeal™ Glass Bottle
Catalog number FSA428701000
Price (MYR)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-33-5, 75-09-2
Specifications
Chemical Name or MaterialIodine monobromide
CAS7789-33-5
ColorRed
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Suspected of causing cancer.