Check Order Status
1-Benzylindole-3-carboxylic acid, 98%, Thermo Scientific™
Element Search
Structure Search
1-Benzylindole-3-carboxylic acid, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Packaging:
Glass bottle
Catalog number
FSA428260010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
1-Benzylindole-3-carboxylic acid
CAS
27018-76-4
CAS Min %
97.5
CAS Max %
100.0
Assay Percent Range
97.5% min. (HPLC)