n-Hexadecyl palmitate, 95%
n-Hexadecyl palmitate, 95%
n-Hexadecyl palmitate, 95%
n-Hexadecyl palmitate, 95%
n-Hexadecyl palmitate, 95%
CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA428222500
Price (MYR)
-
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexadecyl palmitate
CAS540-10-3
Melting Point52°C to 56°C
EINECS Number208-736-6
Boiling Point360°C