(±)-alpha-Tocopherol, 95%, synthetic
(±)-alpha-Tocopherol, 95%, synthetic
(±)-alpha-Tocopherol, 95%, synthetic
(±)-alpha-Tocopherol, 95%, synthetic
Thermo Scientific Chemicals
(±)-alpha-Tocopherol, 95%, synthetic
CAS: 10191-41-0 | C29H50O2 | 430.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA428125000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(±)-α-Tocopherol
Name NoteSynthetic
CAS10191-41-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning