L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%
Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%
CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA427995000
Price (MYR)
834.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
834.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-(+)-Tartaric acid dipotassium salt
CAS6100-19-2
Assay Percent Range97%
Molecular FormulaC8H14K4O13
MDL NumberMFCD00013065