Dimethyl 2,6-pyridinedicarboxylate, 99%
Dimethyl 2,6-pyridinedicarboxylate, 99%
Dimethyl 2,6-pyridinedicarboxylate, 99%
Dimethyl 2,6-pyridinedicarboxylate, 99%
Thermo Scientific Chemicals
Dimethyl 2,6-pyridinedicarboxylate, 99%
CAS: 5453-67-8 | C9H9NO4 | 195.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA427940100
Price (MYR)
456.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl 2, 6-pyridinedicarboxylate
CAS5453-67-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for