3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
Thermo Scientific Chemicals
3,5-Dimethoxybenzyl bromide, 95%
CAS: 877-88-3 | C9H11BrO2 | 231.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4273400505 gGlass bottle
Catalog number FSA427340050
Price (MYR)
838.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
838.00
EA
Add to cart
Specifications
Chemical Name or Material3, 5-Dimethoxybenzyl bromide
CAS877-88-3
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0