3-(2-Bromoethyl)indole, 95%
3-(2-Bromoethyl)indole, 95%
3-(2-Bromoethyl)indole, 95%
3-(2-Bromoethyl)indole, 95%
Thermo Scientific Chemicals
3-(2-Bromoethyl)indole, 95%
CAS: 3389-21-7 | C10H10BrN | 224.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA426910050
Price (MYR)
1,509.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,509.00
EA
Add to cart
Specifications
Chemical Name or Material3-(2-Bromoethyl)indole
CAS3389-21-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0