DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Thermo Scientific Chemicals
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Catalog number FSA426380500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS3483-12-3, 7732-18-5
Specifications
Chemical Name or MaterialDL-1, 4-Dithiothreitol
ColorColorless
Name Note1M Solution in water, For molecular biology
CAS3483-12-3
Health Hazard 1GHS Signal Word: Warning
View more
For biochemistry
RUO – Research Use Only