1-Chlorobutane, 99.8%, for HPLC
1-Chlorobutane, 99.8%, for HPLC
1-Chlorobutane, 99.8%, for HPLC
1-Chlorobutane, 99.8%, for HPLC
Thermo Scientific Chemicals
1-Chlorobutane, 99.8%, for HPLC
CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA425880025
Price (MYR)
1,458.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,458.00
EA
Add to cart
Specifications
Chemical Name or Material1-Chlorobutane
Name NoteFor HPLC
Acidity0.0003 meq/g max.
CAS109-69-3
Absorbance1 cm cell vs HPLC grade water, 0.05 max. at 240nm, 0.01 max. at 250nm, 0.005 max. at 260nm, 0.005 max. at 270nm, 1 cm cell vs HPLC grade water at 0nm
View more
For HPLC
RUO – Research Use Only