Neopentyl bromide, 98%
Neopentyl bromide, 98%
Neopentyl bromide, 98%
Neopentyl bromide, 98%
Thermo Scientific Chemicals
Neopentyl bromide, 98%
CAS: 630-17-1 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA425720050
Price (MYR)
408.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
408.00
EA
Add to cart
Specifications
Chemical Name or MaterialNeopentyl bromide
CAS630-17-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0