Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Sulfuric acid, ACS reagent, 95% solution in water
Thermo Scientific Chemicals
Sulfuric acid, ACS reagent, 95% solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Plastic bottle
Catalog number FSA424525000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Melting Point10.0°C
Boiling Point290.0°C
Health Hazard 1Danger