Starch, ACS reagent, soluble
Starch, ACS reagent, soluble
Starch, ACS reagent, soluble
Starch, ACS reagent, soluble
Thermo Scientific Chemicals
Starch, ACS reagent, soluble
CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA424495000
Price (MYR)
658.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
658.00
BOT
Add to cart
Specifications
Chemical Name or MaterialStarch
ColorWhite
Name NoteSoluble
pH5.0 to 7.0 (2% soln. at 25°C)
CAS9005-84-9