Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent
CAS: 20624-25-3 | C5H10NNaS2·3H2O | 225.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA424311000
Price (MYR)
642.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
642.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiethyldithiocarbamic acid, sodium salt trihydrate
CAS20624-25-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
Health Hazard 3GHS P Statement
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if presen