Potassium oxalate monohydrate, 99%, ACS reagent
Potassium oxalate monohydrate, 99%, ACS reagent
Potassium oxalate monohydrate, 99%, ACS reagent
Potassium oxalate monohydrate, 99%, ACS reagent
Thermo Scientific Chemicals
Potassium oxalate monohydrate, 99%, ACS reagent
CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA424025000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium oxalate monohydrate
CAS6487-48-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.