Molybdic acid, ACS reagent
Molybdic acid, ACS reagent
Molybdic acid, ACS reagent
Molybdic acid, ACS reagent
Thermo Scientific Chemicals
Molybdic acid, ACS reagent
CAS: 7782-91-4 | H2MoO4 | 161.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA423985000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMolybdic acid
Name NoteFor Analysis ACS
CAS7782-91-4
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
Oxidation reagent
RUO – Research Use Only