D-Mannitol, ACS reagent
D-Mannitol, ACS reagent
D-Mannitol, ACS reagent
D-Mannitol, ACS reagent
Thermo Scientific Chemicals
D-Mannitol, ACS reagent
CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA423922500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Mannitol
CAS69-65-8
Melting Point165.0°C to 170.0°C
ColorWhite
EINECS Number200-711-8