Hexanes, 99%, ACS reagent, mixture of isomers
Hexanes, 99%, ACS reagent, mixture of isomers
Hexanes, 99%, ACS reagent, mixture of isomers
Hexanes, 99%, ACS reagent, mixture of isomers
Hexanes, 99%, ACS reagent, mixture of isomers
Thermo Scientific Chemicals
Hexanes, 99%, ACS reagent, mixture of isomers
CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA423760025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexanes
Melting Point-95.0°C
Name NoteMixture of isomers
CAS92112-69-1
Health Hazard 1Danger