Acetic acid, 99.7+%, ACS reagent
Acetic acid, 99.7+%, ACS reagent
Acetic acid, 99.7+%, ACS reagent
Acetic acid, 99.7+%, ACS reagent
Thermo Scientific Chemicals
Acetic acid, 99.7+%, ACS reagent
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA423225000
Price (MYR)
376.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
376.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic Acid
CAS64-19-7
CAS Min %99.7
Health Hazard 1Danger
CAS Max %100.0