Ethyl acetate, 99.5%, for spectroscopy ACS
Ethyl acetate, 99.5%, for spectroscopy ACS
Ethyl acetate, 99.5%, for spectroscopy ACS
Ethyl acetate, 99.5%, for spectroscopy ACS
Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for spectroscopy ACS
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Catalog number FSA423175000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Absorbance0.01 max. at 330nm, 0.01 max. at 400nm, 0.05 max. at 275nm, 0.1 max. at 263nm, 0.5 max. at 257nm, 1 max. at 255nm
CAS141-78-6
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only