Potassium nitrite, 96+%, ACS reagent
Potassium nitrite, 96+%, ACS reagent
Potassium nitrite, 96+%, ACS reagent
Potassium nitrite, 96+%, ACS reagent
Thermo Scientific Chemicals
Potassium nitrite, 96+%, ACS reagent
CAS: 7758-09-0 | KNO2 | 85.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Catalog number FSA423065000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium nitrite
Melting Point387.0°C
Name Note96+%
CAS7758-09-0
ColorWhite to Yellow