Triton™ X-114
Triton™ X-114
Triton™ X-114
Thermo Scientific Chemicals
Triton™ X-114
CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA422365000
Price (MYR)
383.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
383.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriton™ X-114
ColorUndesignated
CAS9002-93-1
Health Hazard 1Danger
Physical FormLiquid