Tri-o-tolylphosphine, 99%
Tri-o-tolylphosphine, 99%
Tri-o-tolylphosphine, 99%
Tri-o-tolylphosphine, 99%
Thermo Scientific Chemicals
Tri-o-tolylphosphine, 99%
CAS: 6163-58-2 | C21H21P | 304.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA422320050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-o-tolylphosphine
CAS6163-58-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee