2,4,6-Triphenyl-s-triazine, 98+%
2,4,6-Triphenyl-s-triazine, 98+%
2,4,6-Triphenyl-s-triazine, 98+%
2,4,6-Triphenyl-s-triazine, 98+%
Thermo Scientific Chemicals
2,4,6-Triphenyl-s-triazine, 98+%
CAS: 493-77-6 | C21H15N3 | 309.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA422190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4, 6-Triphenyl-s-triazine
Name Note99%
CAS493-77-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning