Trichloroisocyanuric acid, 99%
Trichloroisocyanuric acid, 99%
Trichloroisocyanuric acid, 99%
Trichloroisocyanuric acid, 99%
Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%
CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA421542500
Price (MYR)
347.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
347.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrichloroisocyanuric acid
CAS87-90-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
C-X bond formation reagent
RUO – Research Use Only